Learn More
Medchemexpress LLC tert-Butyl 3-(2-iodoethoxy)propanoate | 2296723-16-3 | ≥95.0% | 300.13 | 250 MG
SDP
Catalog No. 5000402837
Encompass_Preferred
tert-Butyl 3-(2-iodoethoxy)propanoate is a PROTAC linker, which refers to the PEG composition. It can be used in the synthesis of PROTAC FAK degrader 1. This product is for research use only and not sold to patients.
  • Protac linker
  • Peg composition
  • Used in the synthesis of PROTAC FAK degrader 1
  • Appearance: liquid
  • Color: colorless to light yellow
Catalog No. 50-004-02837 Supplier Medchemexpress LLC Supplier No. 250MGHY44141
tert-Butyl 3-(2-iodoethoxy)propanoate is a PROTAC linker, which refers to the PEG composition. It can be used in the synthesis of PROTAC FAK degrader 1. This product is for research use only and not sold to patients.
  • Protac linker
  • Peg composition
  • Used in the synthesis of PROTAC FAK degrader 1
  • Appearance: liquid
  • Color: colorless to light yellow