Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl 3-(2-iodoethoxy)propanoate | 2296723-16-3 | ≥95.0% | 300.13 | 5 G
SDP

Catalog No. 5000404212
Encompass_Preferred

tert-Butyl 3-(2-iodoethoxy)propanoate is a PROTAC linker that refers to the PEG composition. It can be used in the synthesis of PROTAC FAK degrader 1.

  • Appearance: liquid
  • Color: colorless to light yellow
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protected from light; in solvent: -80°C for 6 months; -20°C for 1 month (protected from light)

Catalog No. 50-004-04212 Supplier Medchemexpress LLC Supplier No. HY441415G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl 3-(2-iodoethoxy)propanoate is a PROTAC linker that refers to the PEG composition. It can be used in the synthesis of PROTAC FAK degrader 1.

  • Appearance: liquid
  • Color: colorless to light yellow
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protected from light; in solvent: -80°C for 6 months; -20°C for 1 month (protected from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.