Learn More
eMolecules​ 877173-80-3 | tert-Butyl 3-(3-ethoxy-3-oxopropanoyl)piperidine-1-carboxylate | Combi-Blocks | MFCD04115571 | 299.367 | C15H25NO5 | 96.000 | CCOC(=O)CC(=O)C1CCCN(C1)C(=O)OC(C)(C)C | 1g | 232335975
Catalog No. 501549511 Shop All eMolecules Inc. Products
Encompass_Preferred
tert-Butyl 3-(3-ethoxy-3-oxopropanoyl)piperidine-1-carboxylate | Combi-Blocks | 877173-80-3 | MFCD04115571 | 299.367 | C15H25NO5 | 96.000 | CCOC(=O)CC(=O)C1CCCN(C1)C(=O)OC(C)(C)C | 1g | 232335975
Catalog No. 50-154-9511 Supplier eMolecules​ Supplier No. 232335975
tert-Butyl 3-(3-ethoxy-3-oxopropanoyl)piperidine-1-carboxylate | Combi-Blocks | 877173-80-3 | MFCD04115571 | 299.367 | C15H25NO5 | 96.000 | CCOC(=O)CC(=O)C1CCCN(C1)C(=O)OC(C)(C)C | 1g | 232335975