Learn More
Medchemexpress LLC Tert-butyl 3,6,9,12-tetraoxapentadec-14-ynoate | 888010-02-4 | MFCD28122944 | >95.0% | C15H26O6 | 10MG
SDP
Catalog No. 5000260436
Encompass_Preferred
Propargyl-PEG3-OCH2-Boc is a Boc-protected PEG3 linker with a terminal propargyl (alkyne) group, designed for use as a building block in click chemistry and linker synthesis. It enables selective conjugation and orthogonal deprotection steps in multi-step synthetic workflows.
  • Provides a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • Boc protecting group enables orthogonal deprotection in multi-step syntheses.
  • PEG3 spacer imparts hydrophilicity and flexible linker length.
  • Supplied in small pack sizes suitable for medicinal chemistry applications.
  • Characterized by defined molecular weight and formula for reproducible reactions.
Catalog No. 50-002-60436 Supplier Medchemexpress LLC Supplier No. HY14003110MG
Propargyl-PEG3-OCH2-Boc is a Boc-protected PEG3 linker with a terminal propargyl (alkyne) group, designed for use as a building block in click chemistry and linker synthesis. It enables selective conjugation and orthogonal deprotection steps in multi-step synthetic workflows.
  • Provides a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • Boc protecting group enables orthogonal deprotection in multi-step syntheses.
  • PEG3 spacer imparts hydrophilicity and flexible linker length.
  • Supplied in small pack sizes suitable for medicinal chemistry applications.
  • Characterized by defined molecular weight and formula for reproducible reactions.