Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-butyl-(3,6-dihydro-2H-pyran-4-yloxy)dimethylsilane

Catalog No. SYX003891G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003891G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003891G Supplier Sigma Aldrich Supplier No. SYX003891G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reaction of 1-chloro-3-fluorobenzene radical cation with nh3 has been investigated by ft-icr spectrometry
  • It reacts with n-butyllithium to yield bicycloadducts via trapping of intermediate benzynes

Specifications

CAS 953390-79-9
Molecular Formula C11H22O2Si
Linear Formula C11H22O2Si
MDL Number MFCD19443926
Quantity 25 g
Molecular Weight (g/mol) 214.38
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.