Learn More
Medchemexpress LLC Tert-butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate | 1638767-25-5 | MFCD27987307 | 100.0% | 198.27 g/mol | C10H18N2O2 | 5 G
SDP
Catalog No. 5000210918
Encompass_Preferred
tert-Butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate is a Boc-protected 3-aminobicyclo[1.1.1]pentane used as a drug intermediate and laboratory reagent for synthesis of active compounds. It is supplied as a high-purity solid suitable for research-scale organic synthesis.
  • Boc-protected primary amine for downstream deprotection and functionalization.
  • High purity: 99.96% (suitable for synthetic chemistry applications).
  • Molecular formula C10H18N2O2 and molecular weight 198.27 g/mol.
  • Available in multiple pack sizes for flexible scale-up, including 5 G for small-scale synthesis.
  • Solid reagent that facilitates incorporation of bicyclo[1.1.1]pentane motifs into target molecules.
Catalog No. 50-002-10918 Supplier Medchemexpress LLC Supplier No. HYW0544245G
tert-Butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate is a Boc-protected 3-aminobicyclo[1.1.1]pentane used as a drug intermediate and laboratory reagent for synthesis of active compounds. It is supplied as a high-purity solid suitable for research-scale organic synthesis.
  • Boc-protected primary amine for downstream deprotection and functionalization.
  • High purity: 99.96% (suitable for synthetic chemistry applications).
  • Molecular formula C10H18N2O2 and molecular weight 198.27 g/mol.
  • Available in multiple pack sizes for flexible scale-up, including 5 G for small-scale synthesis.
  • Solid reagent that facilitates incorporation of bicyclo[1.1.1]pentane motifs into target molecules.