Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate | 1638767-25-5 | MFCD27987307 | 100.0% | 198.27 g/mol | C10H18N2O2 | 50 G
SDP

Catalog No. 5000210917
Encompass_Preferred

Tert-butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate is a Boc-protected amine used as a synthetic intermediate in medicinal chemistry and drug discovery. It is supplied for research use and is commonly used to introduce a bicyclo[1.1.1]pentane moiety into active compounds.

  • Used as a Boc-protected 3-aminobicyclo[1.1.1]pentane building block.
  • Molecular formula: C10H18N2O2.
  • Molecular weight: approximately 198.27 g/mol.
  • High purity suitable for synthesis and medicinal chemistry applications.
  • Typically supplied as a 50 g pack for research-scale synthesis.

Catalog No. 50-002-10917 Supplier Medchemexpress LLC Supplier No. HYW05442450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tert-butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate is a Boc-protected amine used as a synthetic intermediate in medicinal chemistry and drug discovery. It is supplied for research use and is commonly used to introduce a bicyclo[1.1.1]pentane moiety into active compounds.

  • Used as a Boc-protected 3-aminobicyclo[1.1.1]pentane building block.
  • Molecular formula: C10H18N2O2.
  • Molecular weight: approximately 198.27 g/mol.
  • High purity suitable for synthesis and medicinal chemistry applications.
  • Typically supplied as a 50 g pack for research-scale synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.