Learn More
Medchemexpress LLC tert-Butyl (3-aminopropyl)carbamate | 75178-96-0 | 99.82% | 174.24 | 25 G
SDP
Catalog No. 5000403084
Encompass_Preferred
tert-Butyl (3-aminopropyl)carbamate is a biochemical reagent used as a biological material or organic compound for life science research. It plays a key role in the synthesis of spermidine analogues and the Suzuki reaction.
  • Can be used as a biological material or organic compound
  • Plays a key role in the synthesis of spermidine analogues
  • Important in the Suzuki reaction
  • Appears as a solid below 22°C and a liquid above 22°C
  • Colorless to off-white appearance
Catalog No. 50-004-03084 Supplier Medchemexpress LLC Supplier No. 25GHY40172
tert-Butyl (3-aminopropyl)carbamate is a biochemical reagent used as a biological material or organic compound for life science research. It plays a key role in the synthesis of spermidine analogues and the Suzuki reaction.
  • Can be used as a biological material or organic compound
  • Plays a key role in the synthesis of spermidine analogues
  • Important in the Suzuki reaction
  • Appears as a solid below 22°C and a liquid above 22°C
  • Colorless to off-white appearance