Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl 3-ethynylazetidine-1-carboxylate | 287193-01-5 | MFCD17016080 | 99.8% | 181.23 g/mol | C10H15NO2 | 500 MG
SDP

Catalog No. 5000209831
Encompass_Preferred

Tert-butyl 3-ethynylazetidine-1-carboxylate is a chemical intermediate used in medicinal chemistry for the synthesis of nitrogen-containing active compounds. It is the tert-butyl ester derivative of azetidinecarboxylate and is supplied as a high-purity building block.

  • Chemical formula: C10H15NO2
  • Molecular weight: 181.23 g/mol
  • Cas number: 287193-01-5
  • Purity (GC): 99.8%
  • Typical applications: intermediate for synthesis of active compounds
  • Available pack sizes: 1 mg, 5 mg, 10 mg

Catalog No. 50-002-09831 Supplier Medchemexpress LLC Supplier No. HYW050367500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tert-butyl 3-ethynylazetidine-1-carboxylate is a chemical intermediate used in medicinal chemistry for the synthesis of nitrogen-containing active compounds. It is the tert-butyl ester derivative of azetidinecarboxylate and is supplied as a high-purity building block.

  • Chemical formula: C10H15NO2
  • Molecular weight: 181.23 g/mol
  • Cas number: 287193-01-5
  • Purity (GC): 99.8%
  • Typical applications: intermediate for synthesis of active compounds
  • Available pack sizes: 1 mg, 5 mg, 10 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.