Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl 3-fluoro-4-oxopiperidine-1-carboxylate

Catalog No. SYX000941G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX000941G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX000941G Supplier Sigma Aldrich Supplier No. SYX000941G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-(trimethylsiloxy)cyclohexene undergoes michael addition reaction with methyl vinyl ketone in the presence of dibutyltin bis(triflate).1-(trimethylsiloxy)cyclohexene was used in one-pot synthesis of +÷1,9- octalone
  • It is intermediate to many complex compounds including natural products

Specifications

CAS 211108-50-8
Molecular Formula C10H16FNO3
Linear Formula C10H16FNO3
MDL Number MFCD10566554
Quantity 5 g
Molecular Weight (g/mol) 217.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.