Learn More
Sigma Aldrich tert-Butyl 3-formyl-1H-indole-1-carboxylate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 3-methoxypyridine is a substituted pyridine
- Kinetics of the oxidation of 3-methoxypyridine mediated by sulphate radicals has been investigated by flash photolysis of peroxodisulphate, s2o82-
- Ortho lithiation of 3-methoxypyridine has been studied using mesityllithium as the metalating base.3-methoxypyridine may be employed as a catalyst for the addition reaction of various 1,2-acyclic diones to activated acetylenic esters
Specifications
Specifications
| CAS | 57476-50-3 |
| Molecular Formula | C14H15NO3 |
| Linear Formula | C14H15NO3 |
| MDL Number | MFCD00049230 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 245.27 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.