Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl 3-hydroxypropionate

Catalog No. 902181GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-218-1GF 1 g
90-218-5GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-218-1GF Supplier Sigma Aldrich Supplier No. 902181GF
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tert-butyl 3-hydroxypropionate is a key starting material for the synthesis of wortmannilactone c
  • It can also be used to prepare three-dimensional metal-fullerene frameworks (mffs)

Specifications

CAS 59854-11-4
Density 0.990 g/mL (at 20°C (literature))
Molecular Formula C7H14O3
Linear Formula HOCH2CH2COOC(CH3)3
Refractive Index n20/D 1.424
MDL Number MFCD04974574
Quantity 1 g
Molecular Weight (g/mol) 146.18
Percent Purity ≥98.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.