Learn More
Sigma Aldrich tert-Butyl 3-hydroxypropionate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Tert-butyl 3-hydroxypropionate is a key starting material for the synthesis of wortmannilactone c
- It can also be used to prepare three-dimensional metal-fullerene frameworks (mffs)
Specifications
Specifications
| CAS | 59854-11-4 |
| Density | 0.990 g/mL (at 20°C (literature)) |
| Molecular Formula | C7H14O3 |
| Refractive Index | n20/D 1.424 |
| Linear Formula | HOCH2CH2COOC(CH3)3 |
| MDL Number | MFCD04974574 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 146.18 |
| Percent Purity | ≥98.0% (GC) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.