Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl 3-iodopropylcarbamate | 167479-01-8 | >=97.0% | 285.12 g/mol | C8H16INO2 | 5 G
SDP

Catalog No. 5000210012
Encompass_Preferred

Tert-butyl 3-iodopropylcarbamate is a Boc-protected 3-iodopropyl carbamate used as a building block in organic synthesis and medicinal chemistry to introduce a protected 3-iodopropyl group.

  • High purity (>=97.0% by NMR).
  • Solid form with melting point 37-40 °C.
  • Molecular weight 285.12 g/mol and formula C8H16INO2.
  • Recommended storage at -20 °C under nitrogen to maintain stability.
  • In solvent storage: -80 °C for 6 months, -20 °C for 1 month (stored under nitrogen).
  • Hazardous properties include harmful if swallowed and causes skin and eye irritation; may cause respiratory irritation.

Catalog No. 50-002-10012 Supplier Medchemexpress LLC Supplier No. HYW0538425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tert-butyl 3-iodopropylcarbamate is a Boc-protected 3-iodopropyl carbamate used as a building block in organic synthesis and medicinal chemistry to introduce a protected 3-iodopropyl group.

  • High purity (>=97.0% by NMR).
  • Solid form with melting point 37-40 °C.
  • Molecular weight 285.12 g/mol and formula C8H16INO2.
  • Recommended storage at -20 °C under nitrogen to maintain stability.
  • In solvent storage: -80 °C for 6 months, -20 °C for 1 month (stored under nitrogen).
  • Hazardous properties include harmful if swallowed and causes skin and eye irritation; may cause respiratory irritation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.