Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-butyl (3aR,7aS)-hexahydro-3H-benzo[d][1,2,3]oxathiazole-3-carboxylate 2,2-dioxide

Catalog No. VBP002691G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VBP002691G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. VBP002691G Supplier Sigma Aldrich Supplier No. VBP002691G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Diethylenetriaminepentakis(methylphosphonic acid) (dtpmp) is a multidentate chelating agent
  • Hydrogen peroxide oxidation of dtpmp is reported
  • Dtpmp, a phosphonate, is commonly used as crystallization inhibitors.diethylenetriaminepentakis(methylphosphonic acid) solution is suitable for use in a study to investigate the dynamics and kinetics of sodium sulfate crystallization in the presence of various organophosphonic acids
  • It may be employed as ligand for the characterization of ce3+ complexes by luminescence spectroscopy.multidentate complexing agent

Specifications

Molecular Formula C11H19O5N1S1
Linear Formula C11H19O5N1S1
Quantity 100 mL
Molecular Weight (g/mol) 277.34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.