Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (3R)-3-amino-5-methylhexanoate, 97%, Thermo Scientific™

Catalog No. AC337925000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC337925000 500 mg Glass Bottle
AC337921000 100 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC337925000 Supplier Thermo Scientific Chemicals Supplier No. 337925000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 166023-29-6
Molecular Formula C11H24NO2
Molecular Weight (g/mol) 202.32
MDL Number MFCD01076257
InChI Key WBOPVTYXLFEVGP-SECBINFHSA-O
PubChem CID 2733825
IUPAC Name tert-butyl (3R)-3-amino-5-methylhexanoate
SMILES CC(C)C[C@@H]([NH3+])CC(=O)OC(C)(C)C

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4300 to 1.432
Specific Rotation -15
Synonym tert-butyl 3r-3-amino-5-methylhexanoate, hexanoicacid, 3-amino-5-methyl-, 1,1-dimethylethyl ester, 3r, tert-butyl 3r-3-amino-5-methyl-hexanoate, r-tert-butyl 3-amino-5-methylhexanoate, 3r-3-amino-5-methylhexanoic acid tert-butyl ester, hexanoicacid,3-amino-5-methyl-,1,1-dimethylethyl ester, 3r
Specific Rotation Condition - 15.00 (20.00°C c=1,CHCl3)
Quantity 500 mg
Formula Weight 201.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material tert-Butyl (3R)-3-amino-5-methylhexanoate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.