Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (3S)-3-amino-3-phenylpropanoate, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
500mg
1g
120686-18-2
C13H19NO2
221.3
MFCD00798310
TYYCBAISLMKLMT-NSHDSACASA-N
s-tert-butyl 3-amino-3-phenylpropanoate,tert-butyl 3s-3-amino-3-phenylpropanoate,benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, bs,s-tert-butyl3-amino-3-phenylpropanoate,t-butyl 3s-3-amino-3-phenylpropanoate,s-alpha-2-oxo-2-tert-butoxyethyl benzyl amine,tert-butyl-3s-3-amino-3-phenylpropanoate,benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, bs
7021093
tert-butyl (3S)-3-amino-3-phenylpropanoate
CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N
120686-18-2
C13H19NO2
221.3
MFCD00798310
TYYCBAISLMKLMT-NSHDSACASA-N
s-tert-butyl 3-amino-3-phenylpropanoate,tert-butyl 3s-3-amino-3-phenylpropanoate,benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, bs,s-tert-butyl3-amino-3-phenylpropanoate,t-butyl 3s-3-amino-3-phenylpropanoate,s-alpha-2-oxo-2-tert-butoxyethyl benzyl amine,tert-butyl-3s-3-amino-3-phenylpropanoate,benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, bs
7021093
tert-butyl (3S)-3-amino-3-phenylpropanoate
CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N
Specific Rotation -22
Refractive Index 1.4960 to 1.498
Infrared Spectrum Authentic
Quantity 250mg
Specific Rotation Condition - 22.00 (20.00°C c=1,CHCl3)
Color Colorless to Brown
Packaging Glass Bottle
Assay Percent Range 97%
Formula Weight 221.3
Physical Form Slightly Viscous Liquid
Percent Purity 97%
Chemical Name or Material tert-Butyl (3S)-3-amino-3-phenylpropanoate, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger