Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (3S)-3-amino-5-methylhexanoate, 95%

Catalog No. AC337935000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC337935000 500 mg
AC337931000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC337935000 Supplier Thermo Scientific Chemicals Supplier No. 337935000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 166023-30-9 | C11H23NO2 | 201.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166023-30-9
Molecular Formula C11H23NO2
Molecular Weight (g/mol) 201.31
MDL Number MFCD01076260
InChI Key WBOPVTYXLFEVGP-VIFPVBQESA-N
Synonym tert-butyl 3s-3-amino-5-methylhexanoate, s-tert-butyl 3-amino-5-methylhexanoate, hexanoic acid,3-amino-5-methyl-, 1,1-dimethylethyl ester, 3s, tert-butyl 3s-3-amino-5-methyl-hexanoate, 3s-3-amino-5-methylhexanoic acid tert-butyl ester, hexanoic acid,3-amino-5-methyl-,1,1-dimethylethyl ester, 3s
PubChem CID 7021556
IUPAC Name tert-butyl (3S)-3-amino-5-methylhexanoate
SMILES CC(C)CC(CC(=O)OC(C)(C)C)N

Specifications

Color Colorless to Orange-Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4300 to 1.4350 (20°C, 589nm)
Specific Rotation 12
Quantity 500 mg
Formula Weight 201.31
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material tert-Butyl (3S)-3-amino-5-methylhexanoate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.