Learn More
eMolecules​ 455267-29-5 | TERT-BUTYL 4-(1-AMINOETHYL)PIPERIDINE-1-CARBOXYLATE | MFCD11045798 | 1g
Supplier:  eMolecules​ 599166601
Encompass_Preferred
Medchem Express | 5-Bromouridine | 1g | 789374008 | HY-W009163 | 957-75-5 | 323.099 | C9H11BrN2O6
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool<|a>
Catalog No. 50-214-4244