Learn More
Medchemexpress LLC Tert-butyl 4-[2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindol-5-yl]piperazine-1-carboxylate | 2222114-64-7 | MFCD32705088 | 98.8% | C22H26N4O6 | 10MG
SDP
Catalog No. 5000259258
Encompass_Preferred
Thalidomide-piperazine-Boc is a Boc-protected piperazine derivative of thalidomide used as a synthetic intermediate for assembling E3 ligase ligand-linker conjugates such as PROTACs. It is supplied as a high-purity research reagent with defined solubility and storage recommendations.
  • High purity (98.8%).
  • Boc-protected piperazine for linker attachment.
  • Soluble in DMSO (~41.7 mg/mL) with ultrasonic assistance.
  • Powder stable at -20°C for long-term storage; solvent storage recommendations apply.
  • Intended for research use only.
Catalog No. 50-002-59258 Supplier Medchemexpress LLC Supplier No. HY13498210MG
Thalidomide-piperazine-Boc is a Boc-protected piperazine derivative of thalidomide used as a synthetic intermediate for assembling E3 ligase ligand-linker conjugates such as PROTACs. It is supplied as a high-purity research reagent with defined solubility and storage recommendations.
  • High purity (98.8%).
  • Boc-protected piperazine for linker attachment.
  • Soluble in DMSO (~41.7 mg/mL) with ultrasonic assistance.
  • Powder stable at -20°C for long-term storage; solvent storage recommendations apply.
  • Intended for research use only.