Learn More
Medchemexpress LLC Tert-butyl (4-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)butyl)carbamate | 2093388-52-2 | MFCD32671351 | 99.6% | C22H28N4O6 | 10MG
SDP
Catalog No. 5000260794
Encompass_Preferred
Thalidomide-NH-C4-NH-Boc is a thalidomide-derived E3 ligase ligand-linker conjugate that combines a cereblon-binding thalidomide core with a Boc-protected C4 amine linker for use in PROTAC synthesis and related chemical biology applications requiring targeted E3 ligase recruitment.
  • Boc-protected C4 amine linker enables selective deprotection and conjugation.
  • Cereblon-binding thalidomide core suitable for E3 ligase recruitment.
  • High purity (99.6%) for reliable synthetic and analytical results.
  • Available in multiple sizes for small-scale and larger syntheses.
  • Solid, yellow to green appearance; stable when stored protected from light.
Catalog No. 50-002-60794 Supplier Medchemexpress LLC Supplier No. HY13063910MG
Thalidomide-NH-C4-NH-Boc is a thalidomide-derived E3 ligase ligand-linker conjugate that combines a cereblon-binding thalidomide core with a Boc-protected C4 amine linker for use in PROTAC synthesis and related chemical biology applications requiring targeted E3 ligase recruitment.
  • Boc-protected C4 amine linker enables selective deprotection and conjugation.
  • Cereblon-binding thalidomide core suitable for E3 ligase recruitment.
  • High purity (99.6%) for reliable synthetic and analytical results.
  • Available in multiple sizes for small-scale and larger syntheses.
  • Solid, yellow to green appearance; stable when stored protected from light.