Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-[2-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, 97%, Thermo Scientific™

Catalog No. CC30709DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC30709DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC30709DA Supplier Thermo Scientific Chemicals Supplier No. CC30709DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 179250-28-3
Molecular Formula C16H24N2O3
Molecular Weight (g/mol) 292.379
InChI Key MQZUQGHXDLMAKT-UHFFFAOYSA-N
Synonym 2-4-n-boc-piperazinyl benzyl alcohol, tert-butyl 4-2-hydroxymethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, tert-butyl 4-2-hydroxymethyl phenyl piperazine-1-carboxylate, pubchem11335, 1-boc-4-hydroxymethyl-phenyl-piperazine, 1-boc-4-2-hydroxymethyl-phenyl-piperazine, 1-2-hydroxymethyl-phenyl-piperazine-4-carboxylic, 1-t-butoxycarbonyl-4-2-hydroxymethyl-phenyl-piperazine, 4-2-hydroxymethylphenyl piperazine-1-carboxylic acid t-butyl ester
PubChem CID 7127802
IUPAC Name tert-butyl 4-[2-(hydroxymethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CO

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 292.38
Chemical Name or Material tert-Butyl 4-[2-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.