Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate, 97%, Thermo Scientific™

Catalog No. SPB08457DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB08457DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB08457DA Supplier Thermo Scientific Chemicals Supplier No. SPB08457DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 156185-63-6
Molecular Formula C13H25NO3
Molecular Weight (g/mol) 243.347
InChI Key OXPWHPCCUXESFQ-UHFFFAOYSA-N
Synonym tert-butyl 4-3-hydroxypropyl piperidine-1-carboxylate, tert-butyl 4-3-hydroxypropyl tetrahydro-1 2h-pyridinecarboxylate, 3-1-boc-4-piperidyl-1-propanol, 1-n-boc-4-3'-propanol-piperdine, 3-n-boc-piperidin-4-yl propanol, boc-4-piperidinepropanol, 4-3-hydroxypropyl piperidine, n-boc protected, 1-n-boc-4-3'-propanol-piperidine, 4-3-hydroxy-propyl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-3-hydroxypropyl piperidinecarboxylate
PubChem CID 2800739
IUPAC Name tert-butyl 4-(3-hydroxypropyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)CCCO

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 243.34
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.