Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl (4-(4-amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)butyl)carbamate | 1887095-79-5 | 95.0% | 432.27 | 500 MG
SDP

Catalog No. 5000400874
Encompass_Preferred

tert-Butyl (4-(4-amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)butyl)carbamate is a drug intermediate for the synthesis of various active compounds.

  • Appearance: Solid
  • Shipping: Room temperature in continental US
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).

Catalog No. 50-004-00874 Supplier Medchemexpress LLC Supplier No. HY23483500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl (4-(4-amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)butyl)carbamate is a drug intermediate for the synthesis of various active compounds.

  • Appearance: Solid
  • Shipping: Room temperature in continental US
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.