Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-(4-iodophenyl)tetrahydro-1(2H)-pyrazinecarboxylate, Thermo Scientific™

Catalog No. CC30011DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC30011DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC30011DA Supplier Thermo Scientific Chemicals Supplier No. CC30011DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 151978-66-4
Molecular Formula C15H21IN2O2
Molecular Weight (g/mol) 388.249
MDL Number MFCD06200864
InChI Key ZOWJTZMYSLZILG-UHFFFAOYSA-N
Synonym tert-butyl 4-4-iodophenyl piperazine-1-carboxylate, tert-butyl 4-4-iodophenyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-4-iodophenyl piperazine, 1-piperazinecarboxylicacid, 4-4-iodophenyl-, 1,1-dimethylethyl ester, 4-4-iodo-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-4-iodophenyl piperazine-1-carboxylic acid tert-butyl ester, acmc-1bzfs, 4-4-iodophenyl piperazine, n1-boc protected, 1-tert-butoxycarbonyl-4-4-iodophenyl piperazine, 1-tert-butoxycarbonyl-4 4-iodophenyl piperazine
PubChem CID 2795511
IUPAC Name tert-butyl 4-(4-iodophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 388.24
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(4-iodophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.