Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate, ≥97%, Thermo Scientific™

Catalog No. CC43909CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC43909CB 250 mg Amber Glass Bottle
CC43909DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC43909CB Supplier Thermo Scientific Chemicals Supplier No. CC43909CB
Add to Cart

Chemical Identifiers

CAS 857284-20-9
Molecular Formula C15H23N3O3
Molecular Weight (g/mol) 293.367
MDL Number MFCD08060526
InChI Key QKCHWJYIPFBVRD-UHFFFAOYSA-N
Synonym tert-butyl 4-5-hydroxymethyl pyrid-2-yl piperazine-1-carboxylate, tert-butyl 4-5-hydroxymethyl pyridin-2-yl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-5-hydroxymethyl-2-pyridinyl-, 1,1-dimethylethyl ester, 1-piperazinecarboxylicacid,4-5-hydroxymethyl-2-pyridinyl-,1,1-dimethylethyl ester
PubChem CID 7537576
IUPAC Name tert-butyl 4-[5-(hydroxymethyl)pyridin-2-yl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 293.36
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.