Learn More
Medchemexpress LLC tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate | 571188-59-5 | 99.97% | 278.36 | 10 G
SDP
Catalog No. 5000400661
Encompass_Preferred
tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only. This product is a solid, with a color ranging from white to yellow.
  • Used in the synthesis of active compounds
  • Suitable for research applications
  • Available as a white to yellow solid
  • Should be stored away from moisture and light
Catalog No. 50-004-00661 Supplier Medchemexpress LLC Supplier No. HY3220810G
tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only. This product is a solid, with a color ranging from white to yellow.
  • Used in the synthesis of active compounds
  • Suitable for research applications
  • Available as a white to yellow solid
  • Should be stored away from moisture and light