Learn More
Medchemexpress LLC Tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate | 571188-59-5 | 99.97% | 278.36 | 500 G
SDP
Catalog No. 5000401237
Encompass_Preferred
tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and is not suitable for patient use. The compound appears as a solid with a white to yellow color.
  • For research use only
  • Solid appearance
  • Color ranges from white to yellow
  • Stored at 4°C, sealed, away from moisture and light
Catalog No. 50-004-01237 Supplier Medchemexpress LLC Supplier No. HY32208500G
tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and is not suitable for patient use. The compound appears as a solid with a white to yellow color.
  • For research use only
  • Solid appearance
  • Color ranges from white to yellow
  • Stored at 4°C, sealed, away from moisture and light