Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl 4-formylpiperidine-1-carboxylate | 137076-22-3 | 99.54% | C11H19NO3 | 25 G
SDP

Catalog No. 5000358965
Encompass_Preferred

tert-Butyl 4-formylpiperidine-1-carboxylate is a PROTAC linker used in the synthesis of PROTAC BRM/BRG1 degrader-3 (HY-168251). PROTACs utilize the intracellular ubiquitin-proteasome system for selective protein degradation. This product has a molecular weight of 213.27 and appears as a white to yellow solid below 34°C, becoming liquid above 40°C.

  • Target: PROTAC Linkers
  • Pathway: PROTAC
  • Molecular weight: 213.27
  • Appearance: Solid below 34°C, liquid above 40°C
  • Color: White to yellow
  • Recommended storage temperature: -20°C, stored under nitrogen
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Catalog No. 50-003-58965 Supplier Medchemexpress LLC Supplier No. 25GHY69260
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl 4-formylpiperidine-1-carboxylate is a PROTAC linker used in the synthesis of PROTAC BRM/BRG1 degrader-3 (HY-168251). PROTACs utilize the intracellular ubiquitin-proteasome system for selective protein degradation. This product has a molecular weight of 213.27 and appears as a white to yellow solid below 34°C, becoming liquid above 40°C.

  • Target: PROTAC Linkers
  • Pathway: PROTAC
  • Molecular weight: 213.27
  • Appearance: Solid below 34°C, liquid above 40°C
  • Color: White to yellow
  • Recommended storage temperature: -20°C, stored under nitrogen
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.