Learn More
Medchemexpress LLC tert-Butyl 4-iodopiperidine-1-carboxylate | 301673-14-3 | 311.16 | 1 ML
SDP
Catalog No. 5000445383
Encompass_Preferred
tert-Butyl 4-iodopiperidine-1-carboxylate is a biochemical reagent suitable for life science research. It serves as an organic compound or biological material, for research use only and not intended for patient use.
  • Can be used as a biological material or organic compound
  • Suitable for life science research applications
  • Utilized in coupling reactions for arylation of azetidines, pyrrolidines, and piperidines
Catalog No. 50-004-45383 Supplier Medchemexpress LLC Supplier No. HYW00317710MM/1ML
tert-Butyl 4-iodopiperidine-1-carboxylate is a biochemical reagent suitable for life science research. It serves as an organic compound or biological material, for research use only and not intended for patient use.
  • Can be used as a biological material or organic compound
  • Suitable for life science research applications
  • Utilized in coupling reactions for arylation of azetidines, pyrrolidines, and piperidines