Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl 4-iodopiperidine-1-carboxylate | 301673-14-3 | 311.16 | 10 G
SDP

Catalog No. 5000445739
Encompass_Preferred

tert-Butyl 4-iodopiperidine-1-carboxylate is a biochemical reagent that can be utilized as a biological material or an organic compound for life science-related research. This solid compound has several key features and applications:

  • Biochemical reagent
  • Appearance: solid
  • Color: white to light yellow
  • Molecular weight: 311.16
  • Formula: C10H18INO2
  • CAS no.: 301673-14-3
  • Solubility: soluble in DMSO (100 mg/mL, 321.38 mM)
  • Storage: 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Used in coupling reactions, specifically in iron- and cobalt-catalyzed arylation of azetidines, pyrrolidines, and piperidines with Grignard reagents

Catalog No. 50-004-45739 Supplier Medchemexpress LLC Supplier No. HYW00317710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl 4-iodopiperidine-1-carboxylate is a biochemical reagent that can be utilized as a biological material or an organic compound for life science-related research. This solid compound has several key features and applications:

  • Biochemical reagent
  • Appearance: solid
  • Color: white to light yellow
  • Molecular weight: 311.16
  • Formula: C10H18INO2
  • CAS no.: 301673-14-3
  • Solubility: soluble in DMSO (100 mg/mL, 321.38 mM)
  • Storage: 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Used in coupling reactions, specifically in iron- and cobalt-catalyzed arylation of azetidines, pyrrolidines, and piperidines with Grignard reagents

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.