Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-butyl 5,5-dimethyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide

Catalog No. VBP002681G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VBP002681G 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. VBP002681G Supplier Sigma Aldrich Supplier No. VBP002681G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ammonium lauryl sulfate is an anionic surfactant which can be used in the preparation of porous building ceramics by gelcasting and formulation of cosmetic products
  • It can also be used as a corrosion inhibitor for carbon steel in acidic solution

Specifications

Molecular Formula C9H17O5N1S1
Linear Formula C9H17O5N1S1
MDL Number MFCD29920228
Quantity 250 mL
Molecular Weight (g/mol) 251.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.