Learn More
Medchemexpress LLC tert-Butyl 5-bromoisoindoline-2-carboxylate | 201940-08-1 | 298.18 | 10 G
SDP
Catalog No. 5000447503
Encompass_Preferred
Tert-Butyl 5-bromoisoindoline-2-carboxylate serves as a versatile PROTAC linker, playing a crucial role in synthetic chemistry. It can be used in the synthesis of PTOTAC HSD17B13 degrader 1. This compound is intended strictly for research purposes.
  • Functions as a PROTAC linker
  • Used in the synthesis of PTOTAC HSD17B13 degrader 1
  • Intended for research use only
Catalog No. 50-004-47503 Supplier Medchemexpress LLC Supplier No. HYW00369610G
Tert-Butyl 5-bromoisoindoline-2-carboxylate serves as a versatile PROTAC linker, playing a crucial role in synthetic chemistry. It can be used in the synthesis of PTOTAC HSD17B13 degrader 1. This compound is intended strictly for research purposes.
  • Functions as a PROTAC linker
  • Used in the synthesis of PTOTAC HSD17B13 degrader 1
  • Intended for research use only