Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl 5-bromoisoindoline-2-carboxylate | 201940-08-1 | MFCD09953049 | 298.18 | 5 G
SDP

Catalog No. 5000449247
Encompass_Preferred

Tert-butyl 5-bromoisoindoline-2-carboxylate is a PROTAC Linker, commonly utilized in the synthesis of PTOTAC HSD17B13 degrader 1. This compound has a purity of 99.44% and a molecular weight of 298.18. It appears as a white to off-white solid.

  • PROTAC Linker
  • Used in synthesis of PTOTAC HSD17B13 degrader 1
  • Purity of 99.44%
  • Molecular weight of 298.18
  • White to off-white solid

Catalog No. 50-004-49247 Supplier Medchemexpress LLC Supplier No. HYW0036965G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tert-butyl 5-bromoisoindoline-2-carboxylate is a PROTAC Linker, commonly utilized in the synthesis of PTOTAC HSD17B13 degrader 1. This compound has a purity of 99.44% and a molecular weight of 298.18. It appears as a white to off-white solid.

  • PROTAC Linker
  • Used in synthesis of PTOTAC HSD17B13 degrader 1
  • Purity of 99.44%
  • Molecular weight of 298.18
  • White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.