Learn More
Medchemexpress LLC Tert-butyl (5-hydroxypentyl)carbamate | 75178-90-4 | MFCD01862955 | ≥95.0% | 203.28 g/mol | C10H21NO3 | 5 G
SDP
Catalog No. 5000301196
Encompass_Preferred
Tert-butyl (5-hydroxypentyl)carbamate is a Boc-protected amino alcohol used as a synthetic building block and linker in organic and medicinal chemistry. The molecule combines an acid-labile Boc-protected amine with a terminal hydroxyl, allowing selective deprotection and subsequent functionalization for conjugation or further synthesis.
  • Boc-protected amine removable under mild acidic conditions.
  • Terminal hydroxyl enables esterification, etherification, or activation for coupling.
  • Useful as a bifunctional building block for organic and medicinal chemistry.
  • Molecular weight 203.28 g/mol; formula C10H21NO3.
Catalog No. 50-003-01196 Supplier Medchemexpress LLC Supplier No. HYW0397295G
Tert-butyl (5-hydroxypentyl)carbamate is a Boc-protected amino alcohol used as a synthetic building block and linker in organic and medicinal chemistry. The molecule combines an acid-labile Boc-protected amine with a terminal hydroxyl, allowing selective deprotection and subsequent functionalization for conjugation or further synthesis.
  • Boc-protected amine removable under mild acidic conditions.
  • Terminal hydroxyl enables esterification, etherification, or activation for coupling.
  • Useful as a bifunctional building block for organic and medicinal chemistry.
  • Molecular weight 203.28 g/mol; formula C10H21NO3.