Learn More
Medchemexpress LLC tert-butyl 6-[2-chloro-4-(3-methylimidazol-4-yl)anilino]-2-(1-methylpyrazol-4-yl)pyrrolo[3,2-c]pyrid... | 1400284-80-1 | 99.1% | 503.98 g/mol | C26H26ClN7O2 | 5MG
SDP
Supplier:  Medchemexpress LLC HY126035MG
Encompass_Preferred
CCT251455 is a potent and selective inhibitor of the mitotic kinase monopolar spindle 1 (MPS1/TTK) with an in vitro IC50 of 3 nM, supplied as a solid research-only compound. It shows high selectivity and favourable oral pharmacokinetic profiles in rodent models and is accompanied by a datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-39257