Learn More
Medchemexpress LLC tert-Butyl 6-bromohexanoate | 65868-63-5 | 99.7% | 251.16 | 5 G
SDP
Catalog No. 5000403962
Encompass_Preferred
tert-Butyl 6-bromohexanoate is a PROTAC linker used in chemical research. It serves as a crucial component for designing PROTACs, including PROTAC ERK5 degrader-1.
  • Functions as a PROTAC linker
  • Suitable for designing PROTACs
  • High purity of 99.72%
  • Liquid, colorless to light yellow appearance
  • Specific storage conditions for optimal stability
Catalog No. 50-004-03962 Supplier Medchemexpress LLC Supplier No. HY431915G
tert-Butyl 6-bromohexanoate is a PROTAC linker used in chemical research. It serves as a crucial component for designing PROTACs, including PROTAC ERK5 degrader-1.
  • Functions as a PROTAC linker
  • Suitable for designing PROTACs
  • High purity of 99.72%
  • Liquid, colorless to light yellow appearance
  • Specific storage conditions for optimal stability