Learn More
Medchemexpress LLC tert-Butyl 6-bromohexanoate | 65868-63-5 | 99.7% | 251.16 | 50 G
SDP
Catalog No. 5000404698
Encompass_Preferred
tert-Butyl 6-bromohexanoate is a PROTAC linker. It can be used to design PROTAC, such as PROTAC ERK5 degrader-1.
  • PROTAC linker
  • Can be used to design PROTACs
Catalog No. 50-004-04698 Supplier Medchemexpress LLC Supplier No. HY4319150G
tert-Butyl 6-bromohexanoate is a PROTAC linker. It can be used to design PROTAC, such as PROTAC ERK5 degrader-1.
  • PROTAC linker
  • Can be used to design PROTACs