Learn More
Medchemexpress LLC tert-butyl 7-bromoheptanoate | 51100-47-1 | 97.8% | 265.19 g/mol | C11H21BrO2 | 25 G
SDP
Catalog No. 5000208748
Encompass_Preferred
tert-Butyl 7-bromoheptanoate is an organic building block used as a PROTAC linker in medicinal chemistry and targeted protein degradation research. It provides a brominated alkyl handle for subsequent substitution reactions and is supplied for use in synthesis and linker installation.
  • Used as a PROTAC linker in targeted protein degradation research.
  • Provides a brominated alkyl chain suitable for nucleophilic substitution.
  • Supplied as a liquid, colorless to light yellow.
  • Typical assay around 97.8% for research use.
  • Available in a 25 G pack for synthesis-scale work.
Catalog No. 50-002-08748 Supplier Medchemexpress LLC Supplier No. 25GHYW091879
tert-Butyl 7-bromoheptanoate is an organic building block used as a PROTAC linker in medicinal chemistry and targeted protein degradation research. It provides a brominated alkyl handle for subsequent substitution reactions and is supplied for use in synthesis and linker installation.
  • Used as a PROTAC linker in targeted protein degradation research.
  • Provides a brominated alkyl chain suitable for nucleophilic substitution.
  • Supplied as a liquid, colorless to light yellow.
  • Typical assay around 97.8% for research use.
  • Available in a 25 G pack for synthesis-scale work.