Learn More
Medchemexpress LLC tert-Butyl 8-aminooctanoate | 102522-32-7 | 215.34 | 5 G
SDP
Catalog No. 5000403572
Encompass_Preferred
tert-Butyl 8-aminooctanoate is a PROTAC linker that can be used in the synthesis of PROTACs. It exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E1051 ubiquitin ligase.
  • The other ligand is for the target protein.
Catalog No. 50-004-03572 Supplier Medchemexpress LLC Supplier No. HY427735G
tert-Butyl 8-aminooctanoate is a PROTAC linker that can be used in the synthesis of PROTACs. It exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E1051 ubiquitin ligase.
  • The other ligand is for the target protein.