Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl acetoacetate, 97%

Catalog No. p-7081229
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1111522 100 g
AAA1111536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1111522 Supplier Thermo Scientific Chemicals Supplier No. A1111522
Only null left
Add to Cart

CAS: 1694-31-1 | C8H14O3 | 158.197 g/mol

tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ?,β - unsaturated ketones synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ∞,β - unsaturated ketones synthesis.

Solubility
Soluble in water (9g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1694-31-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00008811
InChI Key JKUYRAMKJLMYLO-UHFFFAOYSA-N
Synonym tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
PubChem CID 15538
IUPAC Name tert-butyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OC(C)(C)C

Specifications

Melting Point -38°C
Density 0.961
pH 5.7
Boiling Point 71°C to 72°C (11 mmHg)
Flash Point 76°C (169°F)
Odor Pleasant
Refractive Index 1.42
Quantity 100 g
Beilstein 1680303
Solubility Information Soluble in water (9g/L).
Formula Weight 158.2
Percent Purity ≥98%
Chemical Name or Material tert-Butyl acetoacetate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 216-904-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.