Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Acetoacetate, MP Biomedicals

Catalog No. ICN209591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209591 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209591 Supplier MP Biomedicals, Inc Supplier No. 0520959180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1694-31-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
InChI Key JKUYRAMKJLMYLO-UHFFFAOYSA-N
Synonym tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
PubChem CID 15538
IUPAC Name tert-butyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OC(C)(C)C

Specifications

Quantity 100 g
Formula Weight 158.2
Chemical Name or Material tert-Butyl Acetoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.