Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl acrylate

Catalog No. 327182100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-718-2100ML 100 mL
32-718-21L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-718-2100ML Supplier Sigma Aldrich Supplier No. 327182100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyl acrylate is a monomer used to produce poly(tert-butyl acrylate) (PtBA) polymers and copolymers, and poly(acrylic acid) polyelectrolyte brushes. It undergoes radiation polymerization to form TBA-gels for radio-fluorogenic dose imaging applications. The monomer can be used to synthesize Poly(t-butyl acrylate). It may be used as a reactive diluent in a study.

Specifications

CAS 1663-39-4
Density 0.875 g/mL (at 25°C)
Boiling Point 61°C to 63°C (60 mmHg)
Molecular Formula C7H12O2
Refractive Index n20/D 1.410 (literature)
Linear Formula CH2=CHCOOC(CH3)3
Quantity 100 mL
MDL Number MFCD00008809
Molecular Weight (g/mol) 128.17
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.