Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl bromoacetate

Catalog No. 12423010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-423-010G 10 g
12-423-0250G 250 g
12-423-050G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-423-010G Supplier Sigma Aldrich Supplier No. 12423010G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tert-butyl bromoacetate serves as building blocks during the synthesis of model n-substituted oligoglycines (peptoids) containing an n-linked lactoside side-chain.tert-butyl bromoacetate has been used in the synthesis of:
  • Nitrilotriacetic acid end-functionalized polystyrene by atom transfer radical polymerization
  • Building block for substituted t-butyl acetates
  • Dihydropyranyl prelinker which is useful in polymer-assisted deprotection of oligosacchararides
  • Collagenase inhibitor (s,s,r)-(-)-actinonin it is the starting reagent for the synthesis of n-oxalylglycine derivatives

Specifications

CAS 5292-43-3
Density 1.321 g/mL (at 25°C (literature))
Boiling Point 50°C (10 mmHg)
Molecular Formula C6H11BrO2
Refractive Index n20/D 1.445 (literature)
Linear Formula BrCH2COOC(CH3)3
MDL Number MFCD00000188
Quantity 10 g
Molecular Weight (g/mol) 195.05
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.