Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl chloride, 98+%

Catalog No. aaa13004ak
Change view
Click to view available options
Quantity:
250 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13004AK 250 mL
AAA13004AP 500 mL
AAA13004AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA13004AK Supplier Thermo Scientific Chemicals Supplier No. A13004AK
Only null left
Add to Cart

CAS: 507-20-0 | C4H9Cl | 92.566 g/mol

tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-20-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000816
InChI Key NBRKLOOSMBRFMH-UHFFFAOYSA-N
Synonym tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
PubChem CID 10486
IUPAC Name 2-chloro-2-methylpropane
SMILES CC(C)(C)Cl

Specifications

Melting Point -26°C
Density 0.846
Boiling Point 50°C to 52°C
Flash Point −27°C (−16°F)
Odor Mild
Refractive Index 1.3848
Quantity 250 mL
UN Number UN1127
Beilstein 1730872
Merck Index 14,1562
Solubility Information Miscible with alcohol,benzene,chloroform and ether. Slightly miscible with water.
Formula Weight 92.57
Percent Purity ≥98%
Chemical Name or Material tert-Butyl chloride
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES
EINECSNumber 208-066-4
RTECSNumber TX5040000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.