Learn More
Medchemexpress LLC Tert-Butyl D-leucinate hydrochloride | 13081-32-8 | ≥97.0% | 223.74 | 100 G
SDP
Catalog No. 5000454720
Encompass_Preferred
tert-Butyl D-leucinate hydrochloride is a leucine derivative that is used as an ergogenic supplement. It influences the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and helps prevent exercise-induced muscle damage. It is recognized as a beneficial ergogenic dietary substance.
  • Leucine derivative
  • Used as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Aids in fuel supply during exercise
  • Improves mental performance during stress
  • Prevents exercise-induced muscle damage
Catalog No. 50-004-54720 Supplier Medchemexpress LLC Supplier No. HYW013686100G
tert-Butyl D-leucinate hydrochloride is a leucine derivative that is used as an ergogenic supplement. It influences the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and helps prevent exercise-induced muscle damage. It is recognized as a beneficial ergogenic dietary substance.
  • Leucine derivative
  • Used as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Aids in fuel supply during exercise
  • Improves mental performance during stress
  • Prevents exercise-induced muscle damage