Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl-DCL | 1025796-31-9 | MFCD28898592 | 99.8% | 487.63 g·mol⁻¹ | C24H45N3O7 | 1 ML
SDP

Catalog No. 5000210914
Encompass_Preferred

Tert-butyl-DCL is a small-molecule inhibitor of prostate-specific membrane antigen (PSMA) supplied as a ready-to-use solution (10 mM in DMSO, 1 mL) with high reported purity used for biochemical and bioimaging research.

  • Ready-to-use 10 mM solution in DMSO.
  • Pack size: 1 mL.
  • High purity (99.8%).
  • CAS number 1025796-31-9 for exact identification.
  • Molecular weight 487.63 g·mol⁻¹; formula C24H45N3O7.

Catalog No. 50-002-10914 Supplier Medchemexpress LLC Supplier No. HYW05429210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Tert-butyl-DCL is a small-molecule inhibitor of prostate-specific membrane antigen (PSMA) supplied as a ready-to-use solution (10 mM in DMSO, 1 mL) with high reported purity used for biochemical and bioimaging research.

  • Ready-to-use 10 mM solution in DMSO.
  • Pack size: 1 mL.
  • High purity (99.8%).
  • CAS number 1025796-31-9 for exact identification.
  • Molecular weight 487.63 g·mol⁻¹; formula C24H45N3O7.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.