Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl-DCL | 1025796-31-9 | MFCD28898592 | 99.8% | 487.63 g/mol | C24H45N3O7 | 250 MG
SDP

Catalog No. 5000209693
Encompass_Preferred

tert-Butyl-DCL is a small-molecule inhibitor of prostate-specific membrane antigen (PSMA) used in cancer research and bioimaging. It exhibits high selectivity and affinity for PSMA and is supplied as a solid or as a DMSO solution in multiple sizes, including 250 mg.

  • High purity (99.8%).
  • Molecular weight 487.63 g/mol.
  • CAS number 1025796-31-9.
  • Available as solid or DMSO solution in multiple sizes, including 250 mg.
  • Useful for PSMA-targeted imaging and related research applications.

Catalog No. 50-002-09693 Supplier Medchemexpress LLC Supplier No. 250MGHYW054292
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tert-Butyl-DCL is a small-molecule inhibitor of prostate-specific membrane antigen (PSMA) used in cancer research and bioimaging. It exhibits high selectivity and affinity for PSMA and is supplied as a solid or as a DMSO solution in multiple sizes, including 250 mg.

  • High purity (99.8%).
  • Molecular weight 487.63 g/mol.
  • CAS number 1025796-31-9.
  • Available as solid or DMSO solution in multiple sizes, including 250 mg.
  • Useful for PSMA-targeted imaging and related research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.