Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tert-Butyl Diazoacetate

Catalog No. 4807541G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-075-41G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 48-075-41G Supplier Sigma Aldrich Supplier No. 4807541G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyl diazoacetate can be used for numerous organic transformations, including cyclopropanation, insertion, and aziridine-forming reactions.-ßtert-Butyl diazoacetate may be used for the preparation of tert-butyl2-(4-chlorophenyl)cyclopropane-1-carboxylate with high enantioselectivity and trans-cis selectivity via cyclopropanation with 4-chlorostyrene in the presence of a chiral (salen)cobalt(III) bromide complex as a catalyst.

Specifications

CAS 63139-21-9
Melting Point 150°C to 155°C
Molecular Formula C8H11BO2
Refractive Index n20/D 1.453
Linear Formula C2H5C6H4B(OH)2
MDL Number MFCD00859377
Quantity 25 g
Synonym (ρ-Ethylphenyl)boronic acid; 4-Ethylbenzeneboronic acid
Molecular Weight (g/mol) 149.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.