Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl ethyl ether, MilliporeSigma™ Supelco™

Catalog No. 111019072 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9072 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9072 Supplier MilliporeSigma Supelco Supplier No. 442795
Add to Cart
Add to Cart

tert-Butyl ethyl ether is a fuel additive used in the formulation of automotive gasoline for reducing the emission of carbon monoxide and unburned hydrocarbons, and also to improve octane ratings.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 637-92-3
Vapor Pressure 155 mmHg (25°C)
Density 0.742 g/mL (at 25°C)
Flash Point -19°C
Physical Form Neat
Linear Formula C2H5 OC(CH3)3
MDL Number MFCD00009225
Melting Point -97°C
Boiling Point 72°C to 73°C
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 102.17
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.